Tisk…

2,3-Dihydro-1-benzofuran-2-ylmethyl-4-methylbenzenesulphonate

Dodavatel: Apollo Scientific

Synonyms: (2,3-Dihydrobenzo[b]furan-2-yl)methyl toluene-4-sulphonate

OR1415-1G OR1415-250MG
APOSOR1415-1GEA 0 CZK
APOSOR1415-1G APOSOR1415-250MG
2,3-Dihydro-1-benzofuran-2-ylmethyl-4-methylbenzenesulphonate
2,3-Dihydro-1-benzofuran-2-ylmethyl-4-methylbenzenesulphonate
Vzorec: C₁₆H₁₆O₄S
MW: 304,37 g/mol
Bod tání: 71,5…73.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06797505
CAS číslo: 94709-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR