Tisk…

Methyl-3-methoxy-4-nitrobenzoate

Dodavatel: Apollo Scientific

OR14144-10G OR14144-1G OR14144-5G
APOSOR14144-10GEA 5430 CZK
APOSOR14144-10G APOSOR14144-1G APOSOR14144-5G
Methyl-3-methoxy-4-nitrobenzoate
Methyl-3-methoxy-4-nitrobenzoate
Vzorec: C₉H₉NO₅
Bod tání: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00051967
CAS číslo: 5081-37-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR