2-Methyl-2H-indazol-5-amine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2-methylindazole, 5-Amino-2-methyl-2H-indazole
OR14143-1G
OR14143-250MG
APOSOR14143-1GEA
1210
CZK
APOSOR14143-1G
APOSOR14143-250MG
2-Methyl-2H-indazol-5-amine
2-Methyl-2H-indazol-5-amine
Vzorec:
C₈H₉N₃ MW: 147,18 g/mol Bod tání: 122…127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03787060 CAS číslo: 60518-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...