Tisk…

tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Dodavatel: Apollo Scientific

OR14124-1G OR14124-5G
APOSOR14124-1GEA 7050 CZK
APOSOR14124-1G APOSOR14124-5G
tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-[2-(3-formylphenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate
Vzorec: C₁₈H₂₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08689721
CAS číslo: 924869-28-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR