1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid, 1-(Ethoxycarbonyl)piperidine-4-carboxylic acid
OR14113-1G
APOSOR14113-1GEA
705
CZK
APOSOR14113-1G
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid
Vzorec:
C₉H₁₅NO₄ MW: 201,22 g/mol Bod tání: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD07339354 CAS číslo: 118133-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...