Tisk…

3-Bromo-1H-pyrrolo[2,3-b]pyridine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromo-7-azaindole

OR14092-1G OR14092-5G
APOSOR14092-1GEA 622 CZK
APOSOR14092-1G APOSOR14092-5G
3-Bromo-1H-pyrrolo[2,3-b]pyridine
3-Bromo-1H-pyrrolo[2,3-b]pyridine
Vzorec: C₇H₅BrN₂
MW: 197,04 g/mol
Bod tání: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD02179276
CAS číslo: 74420-15-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR