Tisk…

2-(Allyloxy)benzaldehyde

Dodavatel: Apollo Scientific

OR14086-25G OR14086-5G
APOSOR14086-25GEA 3310 CZK
APOSOR14086-25G APOSOR14086-5G
2-(Allyloxy)benzaldehyde
2-(Allyloxy)benzaldehyde
Vzorec: C₁₀H₁₀O₂
Bod varu: 92…94 °C/0,04mmHg
Storage Temperature: Ambient
MDL Number: MFCD00014130
CAS číslo: 28752-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR