6-Pyrrolidin-1-ylnicotinaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-(1-Pyrrolidinyl)nicotinaldehyde
OR14012-5G
OR14012-1G
APOSOR14012-5GEA
0
CZK
APOSOR14012-5G
APOSOR14012-1G
6-Pyrrolidin-1-ylnicotinaldehyde
6-Pyrrolidin-1-ylnicotinaldehyde
Vzorec:
C₁₀H₁₂N₂O MW: 176,22 g/mol Bod tání: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200881 CAS číslo: 261715-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...