Tisk…

2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxyquinoline-3-carboxylic acid, 1,2-Dihydro-2-oxoquinoline-3-carboxylic acid

OR14004-1G OR14004-5G
APOSOR14004-1GEA 2820 CZK
APOSOR14004-1G APOSOR14004-5G
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid
Vzorec: C₁₀H₇NO₃
MW: 189,17 g/mol
Bod tání: 330…332 °C
Storage Temperature: Ambient
MDL Number: MFCD01550054
CAS číslo: 2003-79-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR