Tisk…

2-(1H-Pyrazol-1-yl)benzoic acid 97%

Dodavatel: Apollo Scientific

OR1399-1G OR1399-5G OR1399-25G
APOSOR1399-1GEA 2280 CZK
APOSOR1399-1G APOSOR1399-5G APOSOR1399-25G
2-(1H-Pyrazol-1-yl)benzoic acid 97%
2-(1H-Pyrazol-1-yl)benzoic acid
Vzorec: C₁₀H₈N₂O₂
Bod tání: 182,5…183.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03086184
CAS číslo: 55317-53-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR