Tisk…

3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

OR1396-1G OR1396-250MG
APOSOR1396-1GEA 910 CZK
APOSOR1396-1G APOSOR1396-250MG
3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%
3-(Morpholinomethyl)phenylboronic acid pinacol ester
Vzorec: C₁₇H₂₆BNO₃
MW: 303,20 g/mol
Bod tání: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD06797489
CAS číslo: 364794-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR