4,4'-Dimethyl-2,2'-bipyridyl 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4,4'-Dimethyl-2,2'-bipyridine
OR13729-25G
OR13729-5G
OR13729-100G
APOSOR13729-25GEA
1240
CZK
APOSOR13729-25G
APOSOR13729-5G
APOSOR13729-100G
4,4'-Dimethyl-2,2'-bipyridyl 99%
4,4'-Dimethyl-2,2'-bipyridyl
Vzorec:
C₁₂H₁₂N₂ MW: 184,24 g/mol Bod tání: 169…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006441 CAS číslo: 1134-35-6 EINECS: 214-483-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...