Tisk…

6-Chloro-1H-indole-3-carbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Chloroindole-3-carboxaldehyde

OR13728-5G OR13728-1G OR13728-10G
APOSOR13728-5GEA 705 CZK
APOSOR13728-5G APOSOR13728-1G APOSOR13728-10G
6-Chloro-1H-indole-3-carbaldehyde
6-Chloro-1H-indole-3-carbaldehyde
Vzorec: C₉H₆ClNO
MW: 179.60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06657152
CAS číslo: 703-82-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR