3-Bromo-2-methylthiophene
Dodavatel: Apollo Scientific
|
Varování
|
OR13721-1G
APOSOR13721-1GEA
622
CZK
APOSOR13721-1G
3-Bromo-2-methylthiophene
3-Bromo-2-methylthiophene
Vzorec:
C₅H₅BrS MW: 177,0622 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09033008 CAS číslo: 30319-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...