6-Methylnicotinic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid
OR13706-25G
OR13706-1G
APOSOR13706-25GEA
1870
CZK
APOSOR13706-25G
APOSOR13706-1G
6-Methylnicotinic acid 98%
6-Methylnicotinic acid
Vzorec:
C₇H₇NO₂ MW: 137,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006341 CAS číslo: 3222-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...