5-Hydroxy-1H-indazole 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1H-Indazol-5-ol
OR13673-25G
OR13673-5G
OR13673-100G
APOSOR13673-25GEA
2570
CZK
APOSOR13673-25G
APOSOR13673-5G
APOSOR13673-100G
5-Hydroxy-1H-indazole 98%
5-Hydroxy-1H-indazole
Vzorec:
C₇H₆N₂O MW: 134,14 g/mol Bod tání: 186…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD01938124 CAS číslo: 15579-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...