Tisk…

C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Aminomethyl)-2,3-dihydrobenzo[b]furan hydrochloride

OR1366-1G OR1366-5G
APOSOR1366-1GEA 1910 CZK
APOSOR1366-1G APOSOR1366-5G
C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride 97%
C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride
Vzorec: C₉H₁₂ClNO
MW: 185,66 g/mol
Bod tání: 258…259 °C
Storage Temperature: Ambient
MDL Number: MFCD01659957
CAS číslo: 19997-54-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR