Tisk…

Xipamide 95+%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-N-(2,6-dimethylphenyl)-2-hydroxy-5-sulphamoylbenzamide

OR13653-100MG OR13653-250MG
APOSOR13653-100MGEA 4300 CZK
APOSOR13653-100MG APOSOR13653-250MG
Xipamide 95+%
Xipamide
Vzorec: C₁₅H₁₅ClN₂O₄S
MW: 354,81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00865927
CAS číslo: 14293-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR