Pyridiniumchlorid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR13636-100G
OR13636-25G
APOSOR13636-100GEA
1040
CZK
APOSOR13636-100G
APOSOR13636-25G
Pyridiniumchlorid 95%
Pyridinium chloride
Vzorec:
C₅H₆ClN Bod varu: 222…224 °C Bod tání: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012802 CAS číslo: 628-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...