[2-(4-Methyl-1-piperazinyl)phenyl]methanamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR1363-250MG
OR1363-1G
APOSOR1363-250MGEA
1370
CZK
APOSOR1363-250MG
APOSOR1363-1G
[2-(4-Methyl-1-piperazinyl)phenyl]methanamine 97%
[2-(4-Methyl-1-piperazinyl)phenyl]methanamine
Vzorec:
C₁₂H₁₉N₃ MW: 205,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04970994 CAS číslo: 655256-68-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...