3-(Chloromethyl)-5-methylpyridine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR13615-25G
APOSOR13615-25GEA
2940
CZK
APOSOR13615-25G
3-(Chloromethyl)-5-methylpyridine hydrochloride
3-(Chloromethyl)-5-methylpyridine hydrochloride
Vzorec:
C7H9Cl2N MW: 178,0600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11656303 CAS číslo: 1007089-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...