Tisk…

4-Nitropyridin-2-amine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-4-nitropyridine

OR13602-1G OR13602-250MG
APOSOR13602-1GEA 1040 CZK
APOSOR13602-1G APOSOR13602-250MG
4-Nitropyridin-2-amine 98%
4-Nitropyridin-2-amine
Vzorec: C₅H₅N₃O₂
MW: 139,1100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114148
CAS číslo: 4487-50-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR