7-Nitro-1H-indazole 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
OR13550-25G
OR13550-5G
APOSOR13550-25GEA
2900
CZK
APOSOR13550-25G
APOSOR13550-5G
7-Nitro-1H-indazole 97%
7-Nitro-1H-indazole
Vzorec:
C₇H₅N₃O₂ MW: 163,14 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00022789 CAS číslo: 2942-42-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...