Tisk…

4-tert-Butylbenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-tert-Butylbenzene-1-sulphonyl chloride

OR1355-25G OR1355-5G
APOSOR1355-25GEA 1450 CZK
APOSOR1355-25G APOSOR1355-5G
4-tert-Butylbenzenesulphonyl chloride
4-tert-Butylbenzenesulphonyl chloride
Vzorec: C₁₀H₁₃ClO₂S
MW: 232,73 g/mol
Bod tání: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007449
CAS číslo: 15084-51-2
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR