4-Chloropicolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloropyridine-2-carboxylic acid
OR1351-25G
OR1351-5G
APOSOR1351-25GEA
705
CZK
APOSOR1351-25G
APOSOR1351-5G
4-Chloropicolinic acid
4-Chloropicolinic acid
Vzorec:
C₆H₄ClNO₂ MW: 157,56 g/mol Bod tání: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191400 CAS číslo: 5470-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...