Tisk…

8-Methoxyquinoline-5-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR13482-1G OR13482-250MG
APOSOR13482-1GEA 18290 CZK
APOSOR13482-1G APOSOR13482-250MG
8-Methoxyquinoline-5-sulphonyl chloride
8-Methoxyquinoline-5-sulphonyl chloride
Vzorec: C₁₀H₈ClNO₃S
MW: 257.7 g/mol
Bod tání: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD02752892
CAS číslo: 90429-62-2
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR