Tisk…

3-Amino-1H-pyrazole-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-4-pyrazolecarboxylic acid, 3-Aminopyrazole-4-carboxylic acid, 3-Amino-1H-pyrazole-4-carboxylic acid, NSC 89246

OR13449-25G
APOSOR13449-25GEA 4470 CZK
APOSOR13449-25G
3-Amino-1H-pyrazole-4-carboxylic acid
3-Amino-1H-pyrazole-4-carboxylic acid
Vzorec: C₄H₅N₃O₂
MW: 127,10 g/mol
Bod tání: 135 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005239
CAS číslo: 41680-34-6
EINECS: 255-493-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR