Tisk…

[1,2,4]Triazolo[1,5-a]pyrimidine-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR13444-1G OR13444-5G
APOSOR13444-1GEA 2480 CZK
APOSOR13444-1G APOSOR13444-5G
[1,2,4]Triazolo[1,5-a]pyrimidine-2-carboxylic acid
[1,2,4]Triazolo[1,5-a]pyrimidine-2-carboxylic acid
Vzorec: C₆H₄N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00182069
CAS číslo: 202065-25-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR