Tisk…

4-Amino-1,2,5-oxadiazole-3-carbonitrile

Dodavatel: Apollo Scientific
Varování

OR13380-1G OR13380-250MG OR13380-5G
APOSOR13380-1GEA 4040 CZK
APOSOR13380-1G APOSOR13380-250MG APOSOR13380-5G
4-Amino-1,2,5-oxadiazole-3-carbonitrile
4-Amino-1,2,5-oxadiazole-3-carbonitrile
Vzorec: C₃H₂N₄O
Storage Temperature: Ambient
MDL Number: MFCD00235169
CAS číslo: 156463-85-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR