Tisk…

3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-4-methylphenylboronic acid, pinacol ester, HCl

OR1337-1G OR1337-5G
APOSOR1337-1GEA 5460 CZK
APOSOR1337-1G APOSOR1337-5G
3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride 98%
3-Amino-4-methylphenylboronic acid pinacol ester hydrochloride
Vzorec: C₁₃H₂₁BClNO₂
MW: 269,58 g/mol
Bod tání: 234…238 °C
Storage Temperature: Ambient
MDL Number: MFCD06659921
CAS číslo: 850567-52-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR