Tisk…

3-Amino-3-(4-methoxyphenyl)propionic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-3-(4-methoxyphenyl)propionic Acid, 3-Amino-3-(4-methoxyphenyl)propanoic acid

OR13342-1G OR13342-5G
APOSOR13342-1GEA 622 CZK
APOSOR13342-1G APOSOR13342-5G
3-Amino-3-(4-methoxyphenyl)propionic acid
3-Amino-3-(4-methoxyphenyl)propionic acid
Vzorec: C₁₀H₁₃NO₃
MW: 195.2151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187209
CAS číslo: 5678-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR