Tisk…

4-Aminophthalimide 95+%

Dodavatel: Apollo Scientific

Synonyms: 5-Aminoisoindoline-1,3-dione, 5-Amino-isoindole-1,3-dione

OR13335-25G
APOSOR13335-25GEA 2070 CZK
APOSOR13335-25G
4-Aminophthalimide 95+%
4-Aminophthalimide
Vzorec: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041854
CAS číslo: 3676-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR