(±)-4-Chloromandelic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloromandelic acid, (±)-4-Chlorophenylglycolic acid
OR13218-100G
OR13218-25G
OR13218-5G
APOSOR13218-100GEA
1450
CZK
APOSOR13218-100G
APOSOR13218-25G
APOSOR13218-5G
(±)-4-Chloromandelic acid
(±)-4-Chloromandelic acid
Vzorec:
C₈H₇ClO₃ MW: 186.59 Bod varu: 361 Bod tání: 117…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042724 CAS číslo: 492-86-4 EINECS: 207-764-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...