5-[(3-Methylbenzyl)sulphanyl]-1,3,4-thiadiazol-2-ylamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-5-(3-methylbenzylthio)-1,3,4-thiadiazole
OR13144-250MG
OR13144-500MG
APOSOR13144-250MGEA
2070
CZK
APOSOR13144-250MG
APOSOR13144-500MG
5-[(3-Methylbenzyl)sulphanyl]-1,3,4-thiadiazol-2-ylamine
5-[(3-Methylbenzyl)sulphanyl]-1,3,4-thiadiazol-2-ylamine
Vzorec:
C₁₀H₁₁N₃S₂ MW: 237,35 g/mol Bod tání: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00448333 CAS číslo: 299442-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...