Tisk…

3-Bromophenyl phenyl ether (BDE No. 2)

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromodiphenyl ether, 1-Bromo-3-phenoxybenzene

OR13134-1G OR13134-5G OR13134-10G
APOSOR13134-1GEA 953 CZK
APOSOR13134-1G APOSOR13134-5G APOSOR13134-10G
3-Bromophenyl phenyl ether (BDE No. 2)
3-Bromophenyl phenyl ether (BDE No. 2)
Vzorec: C₁₂H₉BrO
MW: 249,1000 g/mol
Bod varu: 156…158 °C/10mmHg
Hustota: 1,3713 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00474520
CAS číslo: 6876-00-2
UN: 3082
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR