1-Cyclohexenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR13115-1G
OR13115-5G
OR13115-25G
APOSOR13115-1GEA
622
CZK
APOSOR13115-1G
APOSOR13115-5G
APOSOR13115-25G
1-Cyclohexenylboronic acid
1-Cyclohexenylboronic acid
Vzorec:
C₆H₁₁BO₂ Bod tání: 165 °C Storage Temperature: Freezer |
MDL Number:
MFCD02179497 CAS číslo: 89490-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...