α-Bromo-2-iodotoulene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
α-Bromo-o-iodotoulene, 2-Iodobenzyl bromide, 1-(Bromomethyl)-2-iodobenzene
OR13072-25G
OR13072-100G
APOSOR13072-25GEA
2400
CZK
APOSOR13072-25G
APOSOR13072-100G
α-Bromo-2-iodotoulene 98%
α-Bromo-2-iodotoulene
Vzorec:
C₇H₆BrI MW: 296,93 g/mol Bod tání: 56…57 °C Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236046 CAS číslo: 40400-13-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...