Tisk…

Methyl-5-bromopicolinate

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

OR13058-5G OR13058-1G
APOSOR13058-5GEA 705 CZK
APOSOR13058-5G APOSOR13058-1G
Methyl-5-bromopicolinate
Methyl-5-bromopicolinate
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
Bod tání: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD04112493
CAS číslo: 29682-15-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR