2,6-Dichlorthiobenzamid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Chlorthiamid
OR13055-1G
OR13055-5G
APOSOR13055-1GEA
1240
CZK
APOSOR13055-1G
APOSOR13055-5G
2,6-Dichlorthiobenzamid 98%
2,6-Dichlorthiobenzamid
Vzorec:
C₇H₅Cl₂NS MW: 206.09 Bod tání: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053523 CAS číslo: 1918-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...