Chromone-3-carboxaldehyde
Dodavatel: Apollo Scientific
Synonyms:
4-Oxo-4H-chromene-3-carbaldehyde
OR1305-100G
APOSOR1305-100GEA
5100
CZK
APOSOR1305-100G
Chromone-3-carboxaldehyde
Chromone-3-carboxaldehyde
Vzorec:
C₁₀H₆O₃ MW: 174,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014667 CAS číslo: 17422-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...