4-Aminopyrimidine-5-carboxylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR13043-1G
OR13043-5G
OR13043-25G
APOSOR13043-1GEA
622
CZK
APOSOR13043-1G
APOSOR13043-5G
APOSOR13043-25G
4-Aminopyrimidine-5-carboxylic acid 98%
4-Aminopyrimidine-5-carboxylic acid
Vzorec:
C₅H₅N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00047383 CAS číslo: 20737-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...