Picolinamidine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR1303-1G
OR1303-25G
OR1303-5G
APOSOR1303-1GEA
827
CZK
APOSOR1303-1G
APOSOR1303-25G
APOSOR1303-5G
Picolinamidine hydrochloride
Picolinamidine hydrochloride
Vzorec:
C₆H₈ClN₃ Bod tání: 145…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052271 CAS číslo: 51285-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...