Tisk…

3-(3-Chlorophenyl)propionic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR13022-25G
APOSOR13022-25GEA 1910 CZK
APOSOR13022-25G
3-(3-Chlorophenyl)propionic acid
3-(3-Chlorophenyl)propionic acid
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Bod tání: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD01310801
CAS číslo: 21640-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR