3-Oxocyclohexanecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR12931-1G
OR12931-5G
APOSOR12931-1GEA
2650
CZK
APOSOR12931-1G
APOSOR12931-5G
3-Oxocyclohexanecarboxylic acid
3-Oxocyclohexanecarboxylic acid
Vzorec:
C₇H₁₀O₃ MW: 142,1525 g/mol Bod tání: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320172 CAS číslo: 16205-98-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...