3-(2-Pyrimidinyloxy)benzenecarbaldehyde
Dodavatel: Apollo Scientific
Synonyms:
3-(Pyrimidin-2-yloxy)benzaldehyde
OR12899-1G
OR12899-100MG
OR12899-250MG
OR12899-500MG
OR12899-2.5G
APOSOR12899-1GEA
5590
CZK
APOSOR12899-1G
APOSOR12899-100MG
APOSOR12899-250MG
APOSOR12899-500MG
APOSOR12899-2.5G
3-(2-Pyrimidinyloxy)benzenecarbaldehyde
3-(2-Pyrimidinyloxy)benzenecarbaldehyde
Vzorec:
C₁₁H₈N₂O₂ MW: 200,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03791149 CAS číslo: 263349-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...