Tisk…

2-Piperidin-4-ylmethylpyridine dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR12878-100MG OR12878-250MG
APOSOR12878-100MGEA 10550 CZK
APOSOR12878-100MG APOSOR12878-250MG
2-Piperidin-4-ylmethylpyridine dihydrochloride
2-Piperidin-4-ylmethylpyridine dihydrochloride
Vzorec: C₁₁H₁₈Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD03840134
CAS číslo: 886886-02-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR