Tisk…

4-Amino-3,5-dimethylpyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,5-Dimethylpyridin-4-amine

OR12853-1G OR12853-250MG
APOSOR12853-1GEA 2530 CZK
APOSOR12853-1G APOSOR12853-250MG
4-Amino-3,5-dimethylpyridine
4-Amino-3,5-dimethylpyridine
Vzorec: C₇H₁₀N₂
MW: 122,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03413657
CAS číslo: 43078-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR