Tisk…

4-(Piperidin-4-ylsulphanyl)-pyridine dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR12826-100MG OR12826-500MG
APOSOR12826-100MGEA 1780 CZK
APOSOR12826-100MG APOSOR12826-500MG
4-(Piperidin-4-ylsulphanyl)-pyridine dihydrochloride
4-(Piperidin-4-ylsulphanyl)-pyridine dihydrochloride
Vzorec: C₁₀H₁₆Cl₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD03840146
CAS číslo: 105283-60-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR