Tisk…

6-Bromoanthranilic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-6-bromobenzoic acid

OR12773-1G OR12773-5G OR12773-25G
APOSOR12773-1GEA 622 CZK
APOSOR12773-1G APOSOR12773-5G APOSOR12773-25G
6-Bromoanthranilic acid 97%
6-Bromoanthranilic acid
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
Bod tání: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD03618455
CAS číslo: 20776-48-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR