Tisk…

2-(4-Methoxyphenoxy)propylamine

Dodavatel: Apollo Scientific
Nebezpečí

OR12731-100MG OR12731-250MG OR12731-1G
APOSOR12731-100MGEA 6950 CZK
APOSOR12731-100MG APOSOR12731-250MG APOSOR12731-1G
2-(4-Methoxyphenoxy)propylamine
2-(4-Methoxyphenoxy)propylamine
Vzorec: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD02684116
CAS číslo: 93750-30-2
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR